Ethyl 2-chloro-3-methoxyisonicotinate ≥97%

Supplier: AOBChem USA

Warning

52093-500MG 52093-1G 52093-25G 52093-5G
77328-618EA 292.52 USD
77328-618 77352-258 77387-076 77376-534
Ethyl 2-chloro-3-methoxyisonicotinate ≥97%
Ethyl 2-chloro-3-methoxyisonicotinate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₃
MW: 215.634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698366
CAS Number: 1214347-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR