6-Chloro-2-fluoro-3-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52078-500MG 52078-1G 52078-5G 52078-25G
77328-616EA 147.29 USD
77328-616 77345-206 77364-950 77387-074
6-Chloro-2-fluoro-3-formylbenzoic acid ≥97%
6-Chloro-2-fluoro-3-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClFO₃
MW: 202.567 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756878
CAS Number: 2386056-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR