3-Fluoro-6-methoxy-2-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

52066-500MG 52066-1G 52066-5G 52066-25G
77328-606EA 234.84 USD
77328-606 77352-250 77364-942 77387-068
3-Fluoro-6-methoxy-2-(methylthio)benzoic acid ≥97%
3-Fluoro-6-methoxy-2-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃S
Storage Temperature: Ambient
MDL Number: MFCD34178614
CAS Number: 2092767-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR