(3-Fluoro-2,6-dimethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

52044-500MG 52044-1G 52044-5G
77338-602EA 503.67 USD
77338-602 77352-248 77376-526
(3-Fluoro-2,6-dimethylphenyl)methanol ≥97%
(3-Fluoro-2,6-dimethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD28551044
CAS Number: 1785108-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR