3-Fluoro-2,6-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52043-500MG 52043-1G 52043-5G
77337-138EA 408.91 USD
77337-138 77352-246 77376-524
3-Fluoro-2,6-dimethylbenzaldehyde ≥97%
3-Fluoro-2,6-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO
Storage Temperature: Ambient
MDL Number: MFCD28545754
CAS Number: 1780701-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR