Ethyl 3-fluoro-2,6-dimethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

52042-500MG 52042-1G 52042-5G
77338-404EA 486.16 USD
77338-404 77352-244 77376-522
Ethyl 3-fluoro-2,6-dimethylbenzoate ≥95%
Ethyl 3-fluoro-2,6-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD28794287
CAS Number: 1806418-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR