2-(Benzyloxy)-1-iodo-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

52037-500MG 52037-1G 52037-5G 52037-25G
77328-604EA 251.32 USD
77328-604 77352-240 77364-940 77387-066
2-(Benzyloxy)-1-iodo-4-methoxybenzene ≥97%
2-(Benzyloxy)-1-iodo-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃IO₂
MW: 340.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756869
CAS Number: 193333-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR