4-Bromo-2-methyl-5-(trifluoromethyl)-2H-indazole ≥97%

Supplier: AOBChem USA

Warning

52032-500MG 52032-1G 52032-5G
77337-972EA 456.29 USD
77337-972 77352-236 77376-518
4-Bromo-2-methyl-5-(trifluoromethyl)-2H-indazole ≥97%
4-Bromo-2-methyl-5-(trifluoromethyl)-2H-indazole

Precaución: for Research Use Only

Formula: C₉H₆BrF₃N₂
MW: 279.057 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530158
CAS Number: 2734775-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR