1-Bromo-5-fluoro-2-(methoxymethoxy)-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

52013-500MG 52013-1G 52013-5G
77338-600EA 503.67 USD
77338-600 77352-232 77376-516
1-Bromo-5-fluoro-2-(methoxymethoxy)-3-methylbenzene ≥97%
1-Bromo-5-fluoro-2-(methoxymethoxy)-3-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂
MW: 249.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530157
CAS Number: 2734778-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR