1-Bromo-2-(difluoromethoxy)-5-fluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

52012-500MG 52012-1G 52012-5G
77339-930EA 682.89 USD
77339-930 77352-230 77376-514
1-Bromo-2-(difluoromethoxy)-5-fluoro-3-methylbenzene ≥97%
1-Bromo-2-(difluoromethoxy)-5-fluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O
MW: 255.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564209
CAS Number: 2168854-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR