2-(Benzyloxy)-1-bromo-5-fluoro-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

52011-500MG 52011-1G 52011-5G
77338-036EA 460.41 USD
77338-036 77352-228 77376-512
2-(Benzyloxy)-1-bromo-5-fluoro-3-methylbenzene ≥95%
2-(Benzyloxy)-1-bromo-5-fluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO
MW: 295.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530156
CAS Number: 2734778-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR