1-Bromo-2-ethoxy-5-fluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

52009-500MG 52009-1G 52009-5G
77336-684EA 391.4 USD
77336-684 77352-226 77376-510
1-Bromo-2-ethoxy-5-fluoro-3-methylbenzene ≥97%
1-Bromo-2-ethoxy-5-fluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530154
CAS Number: 1878507-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR