5-Fluoro-2-methoxy-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52007-500MG 52007-1G 52007-5G
77338-818EA 520.15 USD
77338-818 77352-224 77376-508
5-Fluoro-2-methoxy-3-methylbenzaldehyde ≥97%
5-Fluoro-2-methoxy-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06247545
CAS Number: 883531-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR