Ethyl 5-fluoro-2-methoxy-3-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

52006-500MG 52006-1G 52006-5G
77340-390EA 989.83 USD
77340-390 77352-222 77376-506
Ethyl 5-fluoro-2-methoxy-3-methylbenzoate ≥95%
Ethyl 5-fluoro-2-methoxy-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756866
CAS Number: 2105485-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR