Methyl 5-fluoro-2-methoxy-3-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
52005-500MG
52005-1G
52005-5G
77340-308EA
900.22
USD
77340-308
77352-220
77376-504
Methyl 5-fluoro-2-methoxy-3-methylbenzoate ≥97%
Methyl 5-fluoro-2-methoxy-3-methylbenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁FO₃ MW: 198.191 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34756865 CAS Number: 2111565-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...