5-Fluoro-2-methoxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52004-500MG 52004-1G 52004-5G
77339-364EA 582.98 USD
77339-364 77352-218 77376-502
5-Fluoro-2-methoxy-3-methylbenzoic acid ≥97%
5-Fluoro-2-methoxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22415156
CAS Number: 1427382-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR