Methyl 4-(methylthio)benzo[d][1,3]dioxole-5-carboxylate ≥97%

Supplier: AOBChem USA

Warning

51979-500MG 51979-1G 51979-5G
77339-590EA 602.55 USD
77339-590 77352-212 77376-496
Methyl 4-(methylthio)benzo[d][1,3]dioxole-5-carboxylate ≥97%
Methyl 4-(methylthio)benzo[d][1,3]dioxole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄S
MW: 226.249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756864
CAS Number: 2120936-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR