4-(Methylthio)benzo[d][1,3]dioxole-5-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

51978-500MG 51978-1G 51978-5G
77339-298EA 573.71 USD
77339-298 77352-210 77376-494
4-(Methylthio)benzo[d][1,3]dioxole-5-carboxylic acid ≥97%
4-(Methylthio)benzo[d][1,3]dioxole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈O₄S
MW: 212.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902986
CAS Number: 162506-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR