4-(Methylthio)benzo[d][1,3]dioxole-5-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

51977-500MG 51977-1G 51977-5G
77339-492EA 594.31 USD
77339-492 77352-208 77376-492
4-(Methylthio)benzo[d][1,3]dioxole-5-carbaldehyde ≥97%
4-(Methylthio)benzo[d][1,3]dioxole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₈O₃S
MW: 196.223 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19301974
CAS Number: 212332-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR