(3,4-Difluoro-2-isopropoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

51976-500MG 51976-1G 51976-5G
77337-384EA 413.03 USD
77337-384 77352-206 77376-490
(3,4-Difluoro-2-isopropoxyphenyl)methanol ≥97%
(3,4-Difluoro-2-isopropoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂O₂
MW: 202.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564208
CAS Number: 2140484-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR