1-Bromo-5-chloro-2-fluoro-3-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

51972-500MG 51972-1G 51972-5G
77328-598EA 307.97 USD
77328-598 77352-204 77376-488
1-Bromo-5-chloro-2-fluoro-3-(methoxymethyl)benzene ≥97%
1-Bromo-5-chloro-2-fluoro-3-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₇BrClFO
MW: 253.496 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564207
CAS Number: 2734775-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR