3,4-Difluoro-2-methoxy-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

51957-500MG 51957-1G 51957-25G 51957-5G
77328-594EA 305.91 USD
77328-594 77352-200 77387-060 77376-484
3,4-Difluoro-2-methoxy-5-(methylthio)benzoic acid ≥97%
3,4-Difluoro-2-methoxy-5-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₈F₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD34756861
CAS Number: 2807449-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR