Methyl 4-bromo-2-fluoro-[1,1'-biphenyl]-3-carboxylate ≥95%

Supplier: AOBChem USA

Warning

51946-500MG 51946-1G 51946-5G 51946-25G
77328-592EA 164.8 USD
77328-592 77346-166 77364-934 77387-058
Methyl 4-bromo-2-fluoro-[1,1'-biphenyl]-3-carboxylate ≥95%
Methyl 4-bromo-2-fluoro-[1,1'-biphenyl]-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrFO₂
MW: 309.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564206
CAS Number: 2734777-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR