2,4-Dibromo-6-isopropylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

51925-500MG 51925-1G 51925-5G
77336-806EA 394.49 USD
77336-806 77352-192 77376-476
2,4-Dibromo-6-isopropylbenzoic acid ≥97%
2,4-Dibromo-6-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀Br₂O₂
MW: 321.993 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756855
CAS Number: 2755724-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR