(2,3-Dichloro-6-fluorophenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

51922-500MG 51922-1G 51922-5G 51922-25G
77328-584EA 94.76 USD
77328-584 77342-524 77364-930 77376-474
(2,3-Dichloro-6-fluorophenyl)(thiomorpholino)methanone ≥97%
(2,3-Dichloro-6-fluorophenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀Cl₂FNOS
MW: 294.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564205
CAS Number: 2734775-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR