(2,3-Dichloro-6-fluorophenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

51921-1G 51921-5G 51921-25G
77340-714EA 60.77 USD
77340-714 77364-928 77376-472
(2,3-Dichloro-6-fluorophenyl)(morpholino)methanone ≥97%
(2,3-Dichloro-6-fluorophenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀Cl₂FNO₂
MW: 278.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530150
CAS Number: 2734775-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR