(2,3-Dichloro-6-fluorophenyl)(pyrrolidin-1-yl)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
51919-500MG
51919-1G
51919-5G
51919-25G
77328-582EA
86.52
USD
77328-582
77342-250
77364-924
77376-468
(2,3-Dichloro-6-fluorophenyl)(pyrrolidin-1-yl)methanone ≥97%
(2,3-Dichloro-6-fluorophenyl)(pyrrolidin-1-yl)methanone
Precaución: for Research Use Only
Formula:
C₁₁H₁₀Cl₂FNO MW: 262.108 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34178611 CAS Number: 2643368-23-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...