2,3-Dichloro-N-cyclopropyl-6-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

51917-500MG 51917-1G 51917-5G 51917-25G
77328-580EA 86.52 USD
77328-580 77342-248 77364-922 77376-466
2,3-Dichloro-N-cyclopropyl-6-fluorobenzamide ≥97%
2,3-Dichloro-N-cyclopropyl-6-fluorobenzamide

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂FNO
MW: 248.081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178609
CAS Number: 2643367-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR