2,3-Dichloro-6-fluoro-N,N-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

51916-1G 51916-5G 51916-25G
77341-338EA 82.4 USD
77341-338 77364-920 77376-464
2,3-Dichloro-6-fluoro-N,N-dimethylbenzamide ≥97%
2,3-Dichloro-6-fluoro-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₈Cl₂FNO
MW: 236.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178608
CAS Number: 2643367-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR