2,3-Dichloro-6-fluoro-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

51915-1G 51915-5G 51915-25G
77341-336EA 82.4 USD
77341-336 77364-918 77376-462
2,3-Dichloro-6-fluoro-N-methylbenzamide ≥97%
2,3-Dichloro-6-fluoro-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₆Cl₂FNO
Storage Temperature: Ambient
MDL Number: MFCD34178607
CAS Number: 2643367-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR