1,4-Difluoro-2-methoxy-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

51912-500MG 51912-1G 51912-5G
77338-182EA 468.65 USD
77338-182 77352-190 77376-460
1,4-Difluoro-2-methoxy-3-methylbenzene ≥95%
1,4-Difluoro-2-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18395268
CAS Number: 1261747-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR