(3,6-Difluoro-2-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51911-1G 51911-5G 51911-25G
77341-550EA 92.7 USD
77341-550 77364-916 77376-458
(3,6-Difluoro-2-methoxyphenyl)(methyl)sulfane ≥97%
(3,6-Difluoro-2-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈F₂OS
Storage Temperature: Ambient
MDL Number: MFCD28130666
CAS Number: 1823323-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR