4-Ethoxy-3-isopropylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

51904-500MG 51904-1G 51904-5G 51904-25G
77328-578EA 98.88 USD
77328-578 77342-738 77364-914 77376-456
4-Ethoxy-3-isopropylbenzaldehyde ≥97%
4-Ethoxy-3-isopropylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD06247352
CAS Number: 883537-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR