4-Bromo-1-ethoxy-2-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

51898-500MG 51898-1G 51898-5G 51898-25G
77328-576EA 244.11 USD
77328-576 77352-184 77364-912 77387-052
4-Bromo-1-ethoxy-2-isopropylbenzene ≥97%
4-Bromo-1-ethoxy-2-isopropylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24199188
CAS Number: 24591-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR