Methyl 4-(benzyloxy)-3-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

51895-500MG 51895-1G 51895-5G
77338-336EA 478.95 USD
77338-336 77352-180 77376-448
Methyl 4-(benzyloxy)-3-isopropylbenzoate ≥97%
Methyl 4-(benzyloxy)-3-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₈H₂₀O₃
MW: 284.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564204
CAS Number: 2734776-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR