4-(Benzyloxy)-3-isopropylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

51893-500MG 51893-1G 51893-5G
77335-842EA 367.71 USD
77335-842 77352-176 77376-444
4-(Benzyloxy)-3-isopropylbenzoic acid ≥95%
4-(Benzyloxy)-3-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₃
MW: 270.323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902981
CAS Number: 1000604-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR