1-(Benzyloxy)-4-bromo-2-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

51890-500MG 51890-1G 51890-5G 51890-25G
77328-572EA 184.37 USD
77328-572 77347-216 77364-910 77387-050
1-(Benzyloxy)-4-bromo-2-isopropylbenzene ≥97%
1-(Benzyloxy)-4-bromo-2-isopropylbenzene

Precaución: for Research Use Only

Formula: C₁₆H₁₇BrO
Storage Temperature: Ambient
MDL Number: MFCD24199193
CAS Number: 169247-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR