2,2,2-Trifluoro-N-(5-fluoro-2-methylphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

51880-500MG 51880-1G 51880-5G 51880-25G
77328-568EA 110.21 USD
77328-568 77343-268 77364-906 77376-438
2,2,2-Trifluoro-N-(5-fluoro-2-methylphenyl)acetamide ≥97%
2,2,2-Trifluoro-N-(5-fluoro-2-methylphenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₇F₄NO
MW: 221.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00780951
CAS Number: 340034-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR