N-(3,5-Dichlorophenyl)-2,2,2-trifluoroacetamide ≥97%

Supplier: AOBChem USA

Warning

51855-1G 51855-5G 51855-25G
77341-680EA 99.91 USD
77341-680 77364-900 77376-432
N-(3,5-Dichlorophenyl)-2,2,2-trifluoroacetamide ≥97%
N-(3,5-Dichlorophenyl)-2,2,2-trifluoroacetamide

Precaución: for Research Use Only

Formula: C₈H₄Cl₂F₃NO
MW: 258.025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00755564
CAS Number: 106376-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR