1,5-Dibromo-2-methoxy-3-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

51841-500MG 51841-1G 51841-5G 51841-25G
77328-566EA 114.33 USD
77328-566 77343-384 77364-898 77376-430
1,5-Dibromo-2-methoxy-3-nitrobenzene ≥97%
1,5-Dibromo-2-methoxy-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD24449873
CAS Number: 725241-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR