2'-Ethyl-1,1':4',1''-terphenyl ≥97%

Supplier: AOBChem USA

Warning

51839-500MG 51839-1G 51839-5G
77339-912EA 675.68 USD
77339-912 77352-168 77376-428
2'-Ethyl-1,1':4',1''-terphenyl ≥97%
2'-Ethyl-1,1':4',1''-terphenyl

Precaución: for Research Use Only

Formula: C₂₀H₁₈
Storage Temperature: Ambient
MDL Number: MFCD32902977
CAS Number: 2586127-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR