4-Fluoro-2'-methyl-[1,1'-biphenyl]-3,4'-dicarbaldehyde ≥97%

Supplier: AOBChem USA

Warning

51822-500MG 51822-1G 51822-5G
77338-744EA 513.97 USD
77338-744 77352-162 77376-424
4-Fluoro-2'-methyl-[1,1'-biphenyl]-3,4'-dicarbaldehyde ≥97%
4-Fluoro-2'-methyl-[1,1'-biphenyl]-3,4'-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₁FO₂
MW: 242.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902976
CAS Number: 2586126-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR