4'-Methoxy-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

51819-500MG 51819-1G 51819-5G 51819-25G
77328-562EA 278.1 USD
77328-562 77352-158 77364-894 77387-044
4'-Methoxy-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
4'-Methoxy-2-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23570433
CAS Number: 1509012-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR