4'-Chloro-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

51816-500MG 51816-1G 51816-5G
77336-470EA 388.31 USD
77336-470 77352-156 77376-420
4'-Chloro-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
4'-Chloro-2-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO
MW: 230.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24107840
CAS Number: 1517027-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR