(3-Benzyl-4-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

51791-500MG 51791-1G 51791-5G
77338-866EA 527.36 USD
77338-866 77352-148 77376-416
(3-Benzyl-4-methoxyphenyl)boronic acid ≥97%
(3-Benzyl-4-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅BO₃
MW: 242.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28147755
CAS Number: 475272-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR