Methyl 5-ethoxy-2-fluoro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

51784-500MG 51784-1G 51784-5G 51784-25G
77328-552EA 270.89 USD
77328-552 77352-142 77364-884 77387-034
Methyl 5-ethoxy-2-fluoro-3-methylbenzoate ≥97%
Methyl 5-ethoxy-2-fluoro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530146
CAS Number: 2385809-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR