5-Ethoxy-2-fluoro-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

51783-500MG 51783-1G 51783-5G 51783-25G
77328-550EA 161.71 USD
77328-550 77345-880 77364-882 77387-032
5-Ethoxy-2-fluoro-3-methylbenzoic acid ≥97%
5-Ethoxy-2-fluoro-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564199
CAS Number: 2055398-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR