(5-Ethoxy-2-fluoro-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51782-500MG 51782-1G 51782-5G 51782-25G
77328-548EA 277.07 USD
77328-548 77352-140 77364-880 77387-030
(5-Ethoxy-2-fluoro-3-methylphenyl)(methyl)sulfane ≥97%
(5-Ethoxy-2-fluoro-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₃FOS
Storage Temperature: Ambient
MDL Number: MFCD33404004
CAS Number: 2635937-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR