5-Ethoxy-2-fluoro-1-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

51781-500MG 51781-1G 51781-5G 51781-25G
77328-546EA 178.19 USD
77328-546 77346-652 77364-878 77387-028
5-Ethoxy-2-fluoro-1-iodo-3-methylbenzene ≥97%
5-Ethoxy-2-fluoro-1-iodo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀FIO
MW: 280.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564198
CAS Number: 2090348-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR