1-Bromo-5-ethoxy-2-fluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

51780-500MG 51780-1G 51780-5G 51780-25G
77328-544EA 201.88 USD
77328-544 77352-138 77364-876 77387-026
1-Bromo-5-ethoxy-2-fluoro-3-methylbenzene ≥97%
1-Bromo-5-ethoxy-2-fluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756846
CAS Number: 2386336-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR